Learn More
Medchemexpress LLC IFN-lambda 1/IL-29 50ug
SDP
Catalog No. 5000183028
Encompass_Preferred
IFN-lambda 1 (IL-29) is a member of the Type-III interferon family IFN-lambda 1 signals through a heterodimeric receptor complex comprising IFN receptor 1 (IFNLR1) and IL-10 receptor subunit- (IL-10RB) When IFN-lambda 1 binds with the receptor complex Jak1 and Tyk2 will be activated and leads to subsequent tyrosine phosphorylation of the IFN- R1 and activation of STAT1 and STAT2[1] IFN-lambda 1 modulates immunity in infections and autoimmune diseases[2] IFN-lambda 1/IL-29 Protein Human (HEK293 His) is a recombinant human IFN-lambda 1 (G20-T200) with C-terminal 10 His tag which is produced in HEK293
Catalog No. 50-001-83028 Supplier Medchemexpress LLC Supplier No. HYP7255150UG
IFN-lambda 1 (IL-29) is a member of the Type-III interferon family IFN-lambda 1 signals through a heterodimeric receptor complex comprising IFN receptor 1 (IFNLR1) and IL-10 receptor subunit- (IL-10RB) When IFN-lambda 1 binds with the receptor complex Jak1 and Tyk2 will be activated and leads to subsequent tyrosine phosphorylation of the IFN- R1 and activation of STAT1 and STAT2[1] IFN-lambda 1 modulates immunity in infections and autoimmune diseases[2] IFN-lambda 1/IL-29 Protein Human (HEK293 His) is a recombinant human IFN-lambda 1 (G20-T200) with C-terminal 10 His tag which is produced in HEK293