Learn More
Creative Biomart IFN-Omega 1 (IFNW1)
Supplier:  Creative Biomart THP0253
Encompass_Preferred
IFN-Omega 1 Human Recombinant produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 172 amino acids and having a molecular mass of 19.9kDa. The IFN-Omega 1 is purified by proprietary chromatographic techniques.
Catalog No. 50-208-4304