Learn More
Aviva Systems Biology Corp IFNA13 Antibody - C-terminal region (ARP60732_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60732_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFNA13. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human IFNA13. Alternate names: IFNA1ApplicationWB. Species reactivity: Cow, Dog, Goat, Human, Pig, Sheep
Catalog No. 50-287-144