Learn More
Aviva Systems Biology Corp IFNA5 Antibody - middle region (ARP54652_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54652_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFNA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IFNA5. Alternate names: INFA5, IFN-alphaGApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-61276