Learn More
Cayman Chemical IFNaIFNARIN1hydrochlorI 10mg
SDP
Catalog No. 502990075
Encompass_Preferred
An inhibitor of the IFN-α-IFNAR protein-protein interaction (Kd = 4 µM for binding to IFN-α); inhibits virus-induced increases in the production of IFN-α and IL-12 in BM-pDCs infected with MVA or VSV at 18 µM; reduces increases in IL-12 production induced by stimulation with CpG 2216 or poly(I:C) in the same cells
Catalog No. 50-299-0075 Supplier Cayman Chemical Supplier No. 3096010 MG
An inhibitor of the IFN-α-IFNAR protein-protein interaction (Kd = 4 µM for binding to IFN-α); inhibits virus-induced increases in the production of IFN-α and IL-12 in BM-pDCs infected with MVA or VSV at 18 µM; reduces increases in IL-12 production induced by stimulation with CpG 2216 or poly(I:C) in the same cells