Learn More
ACROBiosystems Recombinant Protein;Human IFN-alpha / beta R2 Protein, Fc Tag;HEK293;1MG;IF2,IFNAR2,IFNARB,IFNABR,IFN-R-2,IFN-alpha/beta receptor 2,IFNR2
Catalog No. 501658691
Encompass_Preferred
Recombinant Protein;Human IFN-alpha / beta R2 Protein, Fc Tag;1MG;Human IFNAR2, Fc Tag (IF2-H5255) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Lys 243 (Accession # NP_997467).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.4 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8691 Supplier ACROBiosystems Supplier No. IF2H52551MG
Recombinant Protein;Human IFN-alpha / beta R2 Protein, Fc Tag;1MG;Human IFNAR2, Fc Tag (IF2-H5255) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Lys 243 (Accession # NP_997467).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.4 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.