Learn More
ACROBiosystems Recombinant Protein;Human IFN-alpha / beta R1 Protein, Fc Tag;HEK293;1MG;IF1,IFNAR1,IFNAR,CRF2-1,IFN-R-1,IFNAR-1
Catalog No. 501658688
Encompass_Preferred
Recombinant Protein;Human IFN-alpha / beta R1 Protein, Fc Tag;1MG;Human IFNAR1, Fc Tag (IF1-H5253) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Lys 436 (Accession # NP_000620).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.8 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-165-8688 Supplier ACROBiosystems Supplier No. IF1H52531MG
Recombinant Protein;Human IFN-alpha / beta R1 Protein, Fc Tag;1MG;Human IFNAR1, Fc Tag (IF1-H5253) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Lys 436 (Accession # NP_000620).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.8 kDa. The protein migrates as 110-120 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.