Learn More
ACROBiosystems Recombinant Protein;Human IFN-alpha / beta R2 Protein;HEK293;1MG;IF2,IFNAR2,IFNARB,IFNABR,IFN-R-2,IFN-alpha/beta receptor 2,IFNR2
Catalog No. 501658690
Encompass_Preferred
Recombinant Protein;Human IFN-alpha / beta R2 Protein;1MG;Human IFNAR2, His Tag (IF2-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Lys 243 (Accession # NP_997467).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8690 Supplier ACROBiosystems Supplier No. IF2H52241MG
Recombinant Protein;Human IFN-alpha / beta R2 Protein;1MG;Human IFNAR2, His Tag (IF2-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Lys 243 (Accession # NP_997467).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.