Learn More
Sino Biological Anti-Interferon alpha/beta receptor 2 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10359MM07TF25
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells. Then conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-222-1925