Learn More
Aviva Systems Biology Corp IFNB1 Antibody - middle region (ARP45228_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45228_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFNB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IFNB1. Alternate names: IFB, IFF, IFNB, MGC96956ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-285-191