Learn More
Sino Biological IFNG / IFN-gamma Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11725RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IFNG (rh IFNG; Catalog#11725-HNAS; NP_000610.2; Met 1-Gln 166). IFNG IgG was purified by human IFNG affinity chromatography.
Catalog No. 50-224-7516