Learn More
Epigentek Inc IFNG Polyclonal Antibody (100 ul)
SDP
Supplier:  Epigentek Inc A51717100
Encompass_Preferred
HRP Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Interferon gamma protein (24-162AA). Tested Applications: ELISA. Reactivity: Human. Alternative Names: Interferon gamma.
Catalog No. 50-119-9629