Learn More
ACROBiosystems Recombinant Protein;Human IFN-gamma R1 / IFNGR1 Protein, Fc Tag;HEK293;1MG;IF1,IFNGR1,CD119,CDw119,IFNGR,IFN-gamma-R1,IFNGR-1
Supplier:  ACROBiosystems IF1H52541MG
Encompass_Preferred
Recombinant Protein;Human IFN-gamma R1 / IFNGR1 Protein, Fc Tag;1MG;Human IFN-gamma R1, Fc Tag (IF1-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession # AAH05333).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.4 kDa. The protein migrates as 55-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8320