Learn More
ACROBiosystems Recombinant Protein;Human IFN-gamma R1 / IFNGR1 Protein;HEK293;200UG;IF1,IFNGR1,CD119,CDw119,IFNGR,IFN-gamma-R1,IFNGR-1
Catalog No. 501658299
Encompass_Preferred
Recombinant Protein;Human IFN-gamma R1 / IFNGR1 Protein;200UG;Human IFN-gamma R1, His Tag (IF1-H5223) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession # AAH05333).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.6 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8299 Supplier ACROBiosystems Supplier No. IF1H5223200UG
Recombinant Protein;Human IFN-gamma R1 / IFNGR1 Protein;200UG;Human IFN-gamma R1, His Tag (IF1-H5223) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession # AAH05333).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.6 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.