Learn More
Sino Biological Recombinant Anti-IFNGR1 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 50705R062A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFNGR1 / CD119 (rM IFNGR1 / CD119; Catalog#50705-M08H; P15261; Met1-Asp253) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-228-2552