Learn More
Cayman Chemical IFNGR1 EXrAClular LIgndb 100ug
SDP
Catalog No. 502947105
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged IFNGR1 expressed in HEK293 cells • Amino acids: 1-245 • MW: 27.3 kDa
Catalog No. 50-294-7105 Supplier Cayman Chemical Supplier No. 32006100 UG
Source: Recombinant human C-terminal His-tagged IFNGR1 expressed in HEK293 cells • Amino acids: 1-245 • MW: 27.3 kDa