Learn More
Sino Biological Recombinant Human IFNGR1 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 10338H02H100
Encompass_Preferred
A DNA sequence encoding the human IFNGR1 (NP_000407.1) (Met1-Gly245) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-5717