Learn More
Biomatik Corporation Ifnlr1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10890
Encompass_Preferred
Ifnlr1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IFNLR1. The Ifnlr1 Polyclonal Antibody has been validated for the following applications: ELISA,WB,IHC,IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interferon Lambda Receptor 1 Protein (21-228Aa). Applications: ELISA,WB,IHC,IF. Target Synonyms: IFNLR1. Accession Number: Q8IU57. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5685