Learn More
Biolegend Brilliant Violet 510™ anti-human IFN-γ
Supplier:  Biolegend 502544
Encompass_Preferred
Brilliant Violet 510™ anti-human IFN-γ [4S.B3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: ICFC; Size: 100 tests
Catalog No. NC1397144