Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemosure™ iFOB Test Kits
SDP

Catalog No. 23800500
Change view
Click to view available options
Includes:
10 tests, 10 tubes, 10 mailers
20 tests, 10 home mailers
20 tests, 20 home mailers
30 tests, 30 tubes, no mailers
30 tests, 50 tubes, 50 mailers
50 tests, 50 tubes, no mailers
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
23-800-500 30 tests, 50 tubes, 50 mailers
23-800-511 20 tests, 10 home mailers
23-800-501 10 tests, 10 tubes, 10 mailers
23-800-512 50 tests, 50 tubes, no mailers
23-800-510 30 tests, 30 tubes, no mailers
23-800-523 20 tests, 20 home mailers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 23-800-500 Supplier Hemosure™ Supplier No. T1CK30
Add to Cart
Add to Cart
This item is not returnable. View return policy

Qualitative, one-step, odorless iFOB test with high sensitivity and specificity

  • CLIA Complexity: Waived
  • CPT Codes: G0328QW (colorectal cancer screening); 82274QW (detection of blood in feces form)
  • Sensitivity: 50ng hHB/mL
  • Specificity: over 96%
  • Useful aid in the diagnosis and therapy of gastrointestinal disorders
  • Patient-friendly for use in physicians' offices, labs, hospitals, clinics, or homes
  • As test sample flows up through absorbent device, the labeled antibody-dye conjugate binds to the hemoglobin in specimen, forming an antibody-antigen complex
  • Results within five minutes
  • Positive results: one band in the control (C) region, one band in the test (T) region
  • Negative results: only one band in the control (C) region
  • Low false positive results
  • Reaction components: citric acid, monoclonal and polyclonal antibody, mouse anti-HB antibodies 1 and 2, and goat anti-mouse IgG antibody

If home kit mailers are not needed, they can be excluded from the order. Please contact your Fisher HealthCare account manager for details.

Order Info

The 10-test kit contains 10 collection bottles and 10 home kit mailers; 30-test kit contains 50 collection bottles and 50 home kit mailers.

Specifications

Content And Storage 30 Fecal occult Blood test cassettes, 50 collection tubes, 50 home kit mailers (each kit includes a mailer, 2 collection Papers, patient instructions and return envelope)
CPT Code G0328QW, 82274QW
Clia Complexity Waived
Type iFOB Test Kit
For Use With (Application) Diagnosis and therapy of gastrointestinal disorders
Includes 30 tests, 50 tubes, 50 mailers
Quantity 30 Pack
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.