Learn More
Apexbio Technology LLC Ifosfamide 3778-73-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063488
Encompass_Preferred
Ifosfamide (CAS 3778-73-2) is an oxazaphosphorine-type alkylating agent structurally analogous to cyclophosphamide As a prodrug it undergoes hepatic activation via cytochrome P450-mediated hydroxylation generating reactive metabolites that form crosslinks within the DNA double strands This alkylation induces DNA damage subsequently inhibiting cell proliferation and promoting apoptosis in rapidly dividing cells Due to these cytostatic and cytotoxic properties Ifosfamide is widely evaluated in oncology research contexts particularly for various hematological malignancies and solid tumors
Catalog No. 50-000-63488 Supplier Apexbio Technology LLC Supplier No. A20975.1
Ifosfamide (CAS 3778-73-2) is an oxazaphosphorine-type alkylating agent structurally analogous to cyclophosphamide As a prodrug it undergoes hepatic activation via cytochrome P450-mediated hydroxylation generating reactive metabolites that form crosslinks within the DNA double strands This alkylation induces DNA damage subsequently inhibiting cell proliferation and promoting apoptosis in rapidly dividing cells Due to these cytostatic and cytotoxic properties Ifosfamide is widely evaluated in oncology research contexts particularly for various hematological malignancies and solid tumors