Learn More
Aviva Systems Biology Corp IFRD1 Antibody - N-terminal region (ARP54584_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54584_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFRD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IFRD1. Alternate names: PC4, TIS7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61248