Learn More
Biomatik Corporation Ifrd1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10785
Encompass_Preferred
Ifrd1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IFRD1. The Ifrd1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interferon-Related Developmental Regulator 1 Protein (1-240Aa). Applications: ELISA, WB, IHC. Target Synonyms: IFRD1. Accession Number: O00458. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7410