Learn More
Medchemexpress LLC HY-136057 10mg Medchemexpress, iFSP1 CAS:150651-39-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13605710MG
Encompass_Preferred
Medchemexpress, HY-136057 10mg iFSP1 CAS:150651-39-1 iFSP1 is a potent, selective and glutathione-independent inhibitor of ferroptosis suppressor protein 1 (FSP1) (AIFM2) with an EC50 of 103 nM. iFSP1 selectively induces ferroptosis in GPX4-knockout cells which overexpressed FSP1. iFSP1 is able to sensitize a variety of human cancer cell lines to the ferroptosis inducer, such as (1S,3R)-RSL3 (HY-100218A). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9009