Learn More
Selleck Chemical LLC iFSP1 5mg
SDP
Supplier:  Selleck Chemical LLC S96635MG
Encompass_Preferred
iFSP1 is a potent, selective and glutathione-independent inhibitor of ferroptosis suppressor protein 1 (FSP1/AIFM2) with EC50 of 103 nM. iFSP1 selectively induces ferroptosis in GPX4-knockout Pfa1 and HT1080 cells that overexpresses FSP1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3485