Learn More
Aviva Systems Biology Corp IFT140 Antibody - N-terminal region (ARP36619_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36619_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFT140. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IFT140. Alternate names: DKFZp564L232, FLJ10306, FLJ30571, KIAA0590, WDTC2, c305C8.4, c380F5.1, gs114ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56215