Learn More
Aviva Systems Biology Corp IFT172 Antibody - N-terminal region (ARP51661_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51661_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFT172. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IFT172. Alternate names: DKFZP434A163, KIAA1179, SLB, osm-1, wimApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60579