Learn More
Aviva Systems Biology Corp IFT74 Antibody - C-terminal region (ARP62026_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62026_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFT74. It was validated on Western Blot. . Alternate names: CCDC2, CMG-1, CMG1, FLJ22621, MGC111562ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-547