Learn More
Aviva Systems Biology Corp IFT88 Antibody - middle region (ARP53658_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53658_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IFT88. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human IFT88. Alternate names: D13S1056E, DAF19, MGC26259, RP11-172H24.2, TG737, TTC10, hTg737ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-867