Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig, λ1, λ2 and λ3 Light Chain, Clone: R26-46, Secondary Antibody, BD

Catalog No. BDB553432 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553432 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553432 Supplier BD Biosciences Supplier No. 553432
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The R26-46 antibody reacts specifically with mouse Igs bearing λ1, λ2, or λ3 light chains. It does not react with κ light chain or heavy chain. Detection of surface immunoglobulin on Ig λ chain-secreting hybridoma cells has bee

Host Species: Rat
Clone: R26-46
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig, λ1, λ2 & λ3 Light Chain
Classification Monoclonal
Clone R26-46
Concentration 0.5mg/mL
Host Species Rat
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.