Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig, λ1 Light Chain, Clone: R11-153, Secondary Antibody, BD

Catalog No. BDB553431 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553431 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553431 Supplier BD Biosciences Supplier No. 553431

missing translation for 'validMainframeMsg'

The R11-153 antibody reacts specifically with mouse Igs bearing λ1 light chain. It does not react with κ, λ2, or λ3 light chain or heavy chain. Detection of surface immunoglobulin on mouse B lymphocytes has been demonstrated wit

Host Species: Rat
Clone: R11-153
Isotype: IgG1, κ
Species Reactivity: Mouse
Immunogen: Pooled Mouse Ig

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig, λ1 Light Chain
Classification Monoclonal
Clone R11-153
Concentration 0.5mg/mL
Conjugate Biotin
Host Species Rat
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.