Learn More
INTACT GENOMICS INC ig 10B Chemically Competent Cells (High Efficiency) 1x96 well plate - 20µl/well
Catalog No. 5000083047
Encompass
Intact Genomics ig 10B chemically competent cells (E. coli) are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. This derivative of DH10B provides the highest efficiency in the industry.Product Includes & Storage: ig 10B competent cells: -80 ºC pUC19 control DNA: -20 ºC Recovery medium: 4 ºC
Catalog No. 50-000-83047 Supplier INTACT GENOMICS INC Supplier No. 101896
 Restricted Item
Add to Cart
Add to Cart
Intact Genomics ig 10B chemically competent cells (E. coli) are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. This derivative of DH10B provides the highest efficiency in the industry.Product Includes & Storage: ig 10B competent cells: -80 ºC pUC19 control DNA: -20 ºC Recovery medium: 4 ºC