Learn More
INTACT GENOMICS INC ig 10B Chemically Competent Cells (High Efficiency) 6x200l
Catalog No. 5000082975
Encompass
Intact Genomics ig 10B chemically competent cells (E. coli) are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. This derivative of 10B provides the highest efficiency in the industry.Product Includes & Storage: ig 10B competent cells: -80 C pUC19 control DNA: -20 C Recovery medium: 4 C
Catalog No. 50-000-82975 Supplier INTACT GENOMICS INC Supplier No. 101424
Intact Genomics ig 10B chemically competent cells (E. coli) are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. This derivative of 10B provides the highest efficiency in the industry.Product Includes & Storage: ig 10B competent cells: -80 C pUC19 control DNA: -20 C Recovery medium: 4 C