Learn More
INTACT GENOMICS INC ig 10B Electrocompetent Cells 12x100µl
Catalog No. 5000082976
Encompass
Intact Genomics ig 10B (DH10B derivative) electrocompetent E. coli cells offer the highest transformation efficiencies of ≥5 x 1010 cfu/µg plasmid DNA which are ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction.Product includes and Storage:ig 10B electroCompetent cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC
Catalog No. 50-000-82976 Supplier INTACT GENOMICS INC Supplier No. 121448
Intact Genomics ig 10B (DH10B derivative) electrocompetent E. coli cells offer the highest transformation efficiencies of ≥5 x 1010 cfu/µg plasmid DNA which are ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction.Product includes and Storage:ig 10B electroCompetent cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC