Learn More
INTACT GENOMICS INC ig 10B Electrocompetent Cells 6x100l
Catalog No. 5000082947
Encompass
Intact Genomics ig 10B (10B derivative) electrocompetent E. coli cells offer the highest transformation efficiencies of 5 x 1010 cfu/g plasmid DNA which are ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction.Product includes and Storage:ig 10B electroCompetent cells: -80 CpUC19 control DNA: -20 CRecovery medium: 4 C
Catalog No. 50-000-82947 Supplier INTACT GENOMICS INC Supplier No. 121424
Intact Genomics ig 10B (10B derivative) electrocompetent E. coli cells offer the highest transformation efficiencies of 5 x 1010 cfu/g plasmid DNA which are ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction.Product includes and Storage:ig 10B electroCompetent cells: -80 CpUC19 control DNA: -20 CRecovery medium: 4 C