Learn More
INTACT GENOMICS INC ig 5-alpha Chemically Competent Cells (High Efficiency) 12x50µl
Catalog No. 5000082797
Encompass
Intact Genomics 5 alpha chemically competent E. coli cells are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning. ig 5 alpha chemically competent cells are at least twice the transformation efficiency of the nearest competitor.Product Includes & Storage:ig 5 alpha Competent Cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC
Catalog No. 50-000-82797 Supplier INTACT GENOMICS INC Supplier No. 103112
Intact Genomics 5 alpha chemically competent E. coli cells are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning. ig 5 alpha chemically competent cells are at least twice the transformation efficiency of the nearest competitor.Product Includes & Storage:ig 5 alpha Competent Cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC