Learn More
INTACT GENOMICS INC ig 5-alpha Chemically Competent Cells (High Efficiency) 6x100µl
Supplier:  INTACT GENOMICS INC 103212
Encompass
Intact Genomics 5 alpha chemically competent E. coli cells are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning. ig 5 alpha chemically competent cells are at least twice the transformation efficiency of the nearest competitor.Product Includes & Storage:ig 5 alpha Competent Cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC
Catalog No. 50-000-82850