Learn More
INTACT GENOMICS INC ig 5-alpha Electrocompetent Cells 6x100µl
Catalog No. 5000082982
Encompass
Intact Genomics 5 alpha Electrocompetent E. coli cells are suitable for high efficiency transformation in a wide variety of routine applications such as plasmid isolation, cloning, and subcloning. Mutations in endA1 and recA1 ensure increased plasmid yield and improved plasmid quality.ig 5-alpha Electrocompetent Cells have the following features: ≥2 x 10^10cfu/µg efficiency with electroporation Greatly increased plasmid yield and quality due to endA1 mutation High-efficiency transformation with plasmids 30 kb in size Blue/white screening of recombinant clones due to lacZΔM15 Ensured insert stability due to recA1 mutationProduct Includes and Storage:ig™ 5 alpha Electrocompetent Cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC
Catalog No. 50-000-82982 Supplier INTACT GENOMICS INC Supplier No. 123424
Intact Genomics 5 alpha Electrocompetent E. coli cells are suitable for high efficiency transformation in a wide variety of routine applications such as plasmid isolation, cloning, and subcloning. Mutations in endA1 and recA1 ensure increased plasmid yield and improved plasmid quality.ig 5-alpha Electrocompetent Cells have the following features: ≥2 x 10^10cfu/µg efficiency with electroporation Greatly increased plasmid yield and quality due to endA1 mutation High-efficiency transformation with plasmids 30 kb in size Blue/white screening of recombinant clones due to lacZΔM15 Ensured insert stability due to recA1 mutationProduct Includes and Storage:ig™ 5 alpha Electrocompetent Cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC