Learn More
INTACT GENOMICS INC IG Autoinduction Medium, 5 x 1L
Catalog No. NC3998436
Encompass
Intact Genomics (IG) Autoinduction Medium promotes high-yield growth of E. coli and high-level expression of T7 promoter-regulated recombinant proteins without time-consuming steps such as monitoring optical density (OD) or adding induction components like IPTG. Unlike traditional LB-IPTG induction systems, IG Autoinduction Medium allows regulated protein expression in any expression system that is solely inducible by IPTG from E. coli strains containing a fully functional lac operon.
Catalog No. NC3998436 Supplier INTACT GENOMICS INC Supplier No. 17225L
Intact Genomics (IG) Autoinduction Medium promotes high-yield growth of E. coli and high-level expression of T7 promoter-regulated recombinant proteins without time-consuming steps such as monitoring optical density (OD) or adding induction components like IPTG. Unlike traditional LB-IPTG induction systems, IG Autoinduction Medium allows regulated protein expression in any expression system that is solely inducible by IPTG from E. coli strains containing a fully functional lac operon.