Learn More
INTACT GENOMICS INC IG Chemically Competent Cells, Autoinduction DE3, 6x50 μL, ≥3.0 x 107 cfu/μg
Catalog No. NC3998318
Encompass
These chemically competent cells are suitable for transformation and routine recombinant protein expression from a vector under an IPTG-inducible T7 promoter. They offer simpler, less hands-on autoinduction, potentially leading to increased yields of expressed cells and protein for a wide range of protein targets at various production scales.
  • Suitable for transformation and routine recombinant protein expression.
  • Optimized for autoinduction, offering simpler, less hands-on protein expression.
  • Can lead to significantly increased yields of expressed cells and protein.
  • Enables expression of toxic genes.
  • Tetracycline resistant at 12.5 μg/mL.
  • Transformation efficiency ≥3.0 x 107 cfu/μg pUC19 DNA.
  • Supplied in tubes for convenience.
Catalog No. NC3998318 Supplier INTACT GENOMICS INC Supplier No. 106506
These chemically competent cells are suitable for transformation and routine recombinant protein expression from a vector under an IPTG-inducible T7 promoter. They offer simpler, less hands-on autoinduction, potentially leading to increased yields of expressed cells and protein for a wide range of protein targets at various production scales.
  • Suitable for transformation and routine recombinant protein expression.
  • Optimized for autoinduction, offering simpler, less hands-on protein expression.
  • Can lead to significantly increased yields of expressed cells and protein.
  • Enables expression of toxic genes.
  • Tetracycline resistant at 12.5 μg/mL.
  • Transformation efficiency ≥3.0 x 107 cfu/μg pUC19 DNA.
  • Supplied in tubes for convenience.