Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse Ig Isotyping Instant ELISA™ Kit

Catalog No. 501125163
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-5163 2 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-5163 Supplier Invitrogen™ Supplier No. 885066022
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Mouse Immunoglobulin Isotyping ELISA qualitatively detects Mouse Ig isotypes in mouse hybridoma cell culture supernatants. Principle of the method Biotinylated monoclonal anti-mouse isotype specific or light chain specific capture antibodies bind to the Streptavidin coated plate. Ig Isotype present in the positive control or sample binds to the specific capture antibodies and is simultaneously bound by the HRP-conjugated detector antibody. Following incubation unbound reagents and sample components are removed during a wash step, and substrate solution reactive with HRP is added to the wells. A colored product is formed in proportion to the amount of soluble mouse Ig (H+L) present in the sample. Rigorous validation Each manufactured lot of this ELISA kit is quality tested. See manual for more information on validation.

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Precoated Microplate well plate coated with Monoclonal Antibodies, HRP-Conjugate and Sample Diluent, lyophilized
Positive controls
Wash Buffer Concentrate (20X)
Substrate Solution fast-Tetramethylbenzidine (TMB)
Adhesive Films
Conjugate HRP
Immunoassay Kit Format Coated ELISA Kit
Product Type ELISA
Sample Type Supernatant
Target Species Mouse
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Kit Contents Pre-coated 96 well Microplate coated with Streptavidin, Biotinylated monoclonal anti-mouse isotype or light chain-specific capture antibodies (Isotype-specific: anti-IgG1, IgG2a, IgG2b, IgG2c, IgA, IgM; Light chain specific: lambda or kappa), HRP-Conjugated Anti-Rat lg (H&L) polyclonal antibody, Sample Diluent, lyophilized, Mouse Positive Control: Mouse IgG1, IgG2a, IgG2b, IgG2c, IgA, IgM, kappa and lambda isotype control mix (lyophilized), Wash Buffer Concentrate 20x (PBS with 1% Tween-20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Plate Covers, Biotinylated monoclonal anti-mouse isotype or light chain-specific capture antibodies (Isotype-specific: anti-IgG1, IgG2a, IgG2b, IgG2c, IgA, IgM; Light chain specific: lambda or kappa), HRP-Conjugated Anti-Rat lg (H&L) polyclonal antibody, Sample Diluent, lyophilized, Mouse Positive Control: Mouse IgG1, IgG2a, IgG2b, IgG2c, IgA, IgM, kappa and lambda isotype control mix (lyophilized), Wash Buffer Concentrate 20x (PBS with 1% Tween-20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Plate Covers
Label or Dye Biotin
Quantity 2 x 96 Tests
Regulatory Status RUO
Sample Volume Supernatant 10 μL
Storage Requirements -20°C
Target Mouse Ig
Test Time 30 min
Total Assay Time 2 hr 15 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.