Learn More
Cayman Chemical Ig LIght ChaInhumn RabbIt 50ug
SDP
Catalog No. 502947204
Encompass_Preferred
Immunogen: Human IgG • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human Igĸ, Igλ (-) Cynomolgus monkey, goat, mouse, rat, rhesus monkey IgG • Applications: ELISA, ICC, IHC
Catalog No. 50-294-7204 Supplier Cayman Chemical Supplier No. 3211250 UG
Immunogen: Human IgG • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human Igĸ, Igλ (-) Cynomolgus monkey, goat, mouse, rat, rhesus monkey IgG • Applications: ELISA, ICC, IHC