Learn More
INTACT GENOMICS INC ig Stable 2 Chemically Competent Cells 12x50μl
Supplier:  INTACT GENOMICS INC 101612
Encompass
Intact Genomics (ig) Stable 2 Chemically Competent E. coli cells are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning. Stable 2 cells are capable of cloning methylated genomic sequences, retroviral sequences and direct repeat sequences. Intact Genomics Stable 2 cells provide superb transformation efficiency, allowing for increased opportunity for experimental success.ig Stable 2 Chemically Competent Cells have the following features: • Stable 2 allows for cloning of methylated genomic sequences • Stabilizes retroviral and direct repeat sequences including HIV • High transformation efficiency allows aids in cloning rare sequences • May be used for plasmids > 20 kb • endA1 mutation increases plasmid yield significantly
Catalog No. 50-000-82908