Learn More
INTACT GENOMICS INC ig XL1 Blue Max ElectroCompetent Cells 12x50µl
Catalog No. 5000082981
Encompass
Intact Genomics (ig) XL1 Blue Max Electrocompetent E. coli cells offer the highest transformation efficiency ≥5 x 10^10 cfu/µg plasmid DNA which is ideal for applications such as cDNA or gDNA library construction. These cells can be used to prepare high-quality plasmid DNA, and single strand rescue of phagemid DNA and Blue/white screening. igXL1 Blue Max cells provide transformation efficiencies that are significantly higher than any competitor’s similar product.ig XL1 Blue Max Electrocompetent Cells features: - tetracycline resistant. - endonuclease (endA) deficient, which greatly improves the quality of miniprep DNA. - recombination (recA) deficient, improving insert stability. Cleavage of cloned DNA by the EcoK endonuclease system is prevented by the hsdR mutation. Bue-white color screening via the lacIq Z∆M15 gene on the F´ episome.Product Includes:ig XL1 Blue Max Electrocompetent CellspUC19 control DNARecovery medium
Catalog No. 50-000-82981 Supplier INTACT GENOMICS INC Supplier No. 122324
Intact Genomics (ig) XL1 Blue Max Electrocompetent E. coli cells offer the highest transformation efficiency ≥5 x 10^10 cfu/µg plasmid DNA which is ideal for applications such as cDNA or gDNA library construction. These cells can be used to prepare high-quality plasmid DNA, and single strand rescue of phagemid DNA and Blue/white screening. igXL1 Blue Max cells provide transformation efficiencies that are significantly higher than any competitor’s similar product.ig XL1 Blue Max Electrocompetent Cells features: - tetracycline resistant. - endonuclease (endA) deficient, which greatly improves the quality of miniprep DNA. - recombination (recA) deficient, improving insert stability. Cleavage of cloned DNA by the EcoK endonuclease system is prevented by the hsdR mutation. Bue-white color screening via the lacIq Z∆M15 gene on the F´ episome.Product Includes:ig XL1 Blue Max Electrocompetent CellspUC19 control DNARecovery medium