Learn More
Alpco Ltd IgA (Chicken) ELISA
SDP
Supplier:  Alpco Ltd 41IGACHE01
Encompass
The total Chicken IgA test kits are a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgA in biological samples of Chickens. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-305-141