Learn More
Diagnostic Biosystems IGA-FITC IgA-FITC
Catalog No. 502407619
Encompass_Preferred
This antibody reacts with the alpha chain of human IgA. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.
Catalog No. 50-240-7619 Supplier Diagnostic Biosystems Supplier No. F007
This antibody reacts with the alpha chain of human IgA. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.