Learn More
Athens Research And Technology Inc IGA HUMAN PLASMA 1 MG
Supplier:  Athens Research And Technology Inc 16160907011MG
Encompass_Preferred
Immunoglobulin A, Human Plasma: 1 MG; Frozen in 100 mM Tris-HCl, pH 8.0, with 100 mM NaCl & 0.05% NaN3; MW 170,000; EC=1.32; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-761