Learn More
BioVendor, LLC IgA Secretory Human ELISA (Stool, Saliva), 96 wells
SDP
Supplier:  BioVendor, LLC IC6100
Encompass
IgA Secretory Human ELISA (Stool, Saliva), 96 wells. Enzyme immunoassay kit to measure Immunoglobulin A (Secretory) in human stool or saliva. Quality controls included. SAMPLE REQUIREMENT: 100 l/well. CALIBRATION RANGE: 600-22.2 ng/ml. ASSAY RANGE: 0.5330000 ng/mL. DETECTION LIMIT: 3.1 ng/ml. SUMMARY: sIgA is comprised of two IgA molecules, a J-protein, and a secretory component. The secretory component is synthesized by epithelial cells of the mucous membrane of gastrointestinal, respiratory and urogenital tract. It is also produced by the saliva, tear and mammary glands. sIgA determination allows a first overview of the functionality of the gastrointestinal associated immune system (GALT), giving insight to the secretory power and the degree of stimulation of plasma cells of the intestinal submucosa. sIgA quantification is useful to research allergic disease, infection susceptibility, gut inflammation, autoimmune diseases, etc. Manufactured by Immuchrom GmbH, Germany.
Catalog No. 50-415-8