Learn More
Cayman Chemical IgA1 nd IgA2humn RabbIt 100ug
SDP
Catalog No. 502946564
Encompass_Preferred
Immunogen: Human IgA • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human IgA1, IgA2; (-) Human IgG, IgM, IgD, IgE • Applications: ELISA, ICC, IHC
Catalog No. 50-294-6564 Supplier Cayman Chemical Supplier No. 32114100 UG
Immunogen: Human IgA • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human IgA1, IgA2; (-) Human IgG, IgM, IgD, IgE • Applications: ELISA, ICC, IHC