Learn More
Cayman Chemical IgA2humn RabbIt Monclona 50ug
SDP
Catalog No. 502946372
Encompass_Preferred
Immunogen: Human IgA2 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgA2; (-) Human IgA1, IgG, IgM, IgD, IgE • Applications: ELISA, ICC, IHC
Catalog No. 50-294-6372 Supplier Cayman Chemical Supplier No. 3237050 UG
Immunogen: Human IgA2 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgA2; (-) Human IgA1, IgG, IgM, IgD, IgE • Applications: ELISA, ICC, IHC